Learn More
Aalborg™ Water Direct Glass Flow Tube
Aalborg™ Water Direct Glass Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
Specifications
Type Water Direct Flow Tube
View More Specs
Description
Description
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Specifications
Water Direct Flow Tube
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Aalborg™ Water Direct Glass Flow Tube