missing translation for 'onlineSavingsMsg'
Learn More
Water, Extra Pure, Deionized, Thermo Scientific Chemicals Product Code.: 10347071
In Stock
to see stock.
Add to basket

Water, Extra Pure, Deionized, Thermo Scientific Chemicals

Product Code. 10347071
5 L, Plastic Drum
Click to view available options
Quantity:
5 L
25 L
Packaging:
Plastic Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10347071

Brand: Thermo Scientific Acros 345470050

In Stock
to see stock.
Edge
Add to basket
Add to basket
Request Bulk or Custom Formats
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100.0°C
Conductivity 5.0μS/cm Max.
Linear Formula H2O
Fieser 16,383
Merck Index 15, 10237
Formula Weight 18.02
Grade Extra Pure
Packaging Plastic Drum
Residue after Evaporation 25ppm max.
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 5 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467359 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number345470050,345470250
Lot NumberA0460819 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog Number345470050,345470250
Lot NumberA0460820 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number345470050,345470250
Lot NumberA0458864 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number345470050,345470250
Lot NumberA0449204 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number345470050,345470250
Product Identifier
  • Water
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

  • Multiple Manufacturers
    Save on thousands of eligible products.
    Promo Code: 24111

    Expires2025-03-21

    New Offer
    Save Now

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, Extra Pure, Deionized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.