missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Water For Injection (WFI) for Cell Culture
GREENER_CHOICE

Product Code. 10282830
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
Unit Size:
5000mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10282830

Brand: Gibco™ A1287302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.

  • Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
  • The water is prepared by distillation and filtration (0.1 μm), with no added substances
  • In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
  • Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
  • It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Should be stored at 15° to 30°C
TRUSTED_SUSTAINABILITY

Specifications

pH 4 to 6
Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) None
Quantity 1000 mL
Purification Method Distilled and filtered (0.1 μm)
Shipping Condition Room Temperature
Product Line Gibco

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.