Learn More
Gibco™ Water For Injection (WFI) for Cell Culture
High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.
Brand: Gibco™ A1287303
132.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : CAS Number : 7732-18-5 Weight : 1.00000kg
Description
- Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
- The water is prepared by distillation and filtration (0.1 μm), with no added substances
- In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
- Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
- It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Should be stored at 15° to 30°C
Specifications
4 to 6 | |
Sterile-filtered | |
10 x 500 mL | |
Room Temperature | |
Sustainable packaging |
Cell Culture | |
None | |
Distilled and filtered (0.1 μm) | |
Gibco™ |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.