missing translation for 'onlineSavingsMsg'
Learn More
Water, HPLC for Gradient Analysis, Fisher Chemical™ Product Code.: 10449380

Water, HPLC for Gradient Analysis, Fisher Chemical™

Product Code. 10449380
2.5 L, Amber Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10449380

Brand: Fisher Chemical W/0106/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

HPLC for Gradient Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear
Boiling Point 212°F
Grade HPLC
Packaging Amber Glass Bottle
Quantity 2.5 L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528146 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberW/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS
Lot Number2527436 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberW/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS
Lot Number2526366 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberW/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS
Lot Number2525710 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog NumberW/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS
Product Identifier
  • Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, HPLC for Gradient Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.