missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, J.T. Baker™
32.68 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
1 L
Description
- Particulate Matter: max. 0.0001%
- Residue after Evaporation: max. 2 ppm
- Ultraviolet absorbance of largest eluted peak:
- at 220 nm - max. 0.005 au
- at 254 nm - max. 0.001 au
Specifications
CAS | 7732-18-5 |
Melting Point | 0°C |
Molecular Formula | H2O |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
SMILES | O |
IUPAC Name | water |
Molecular Weight (g/mol) | 18.02 |
Residue after Evaporation | max. 2 ppm |
Quantity | 1 L |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'