missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™

Product Code. 15605360
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
4 x 4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Absorbance max. 0.022 at 210 nm, max. 0.004 at 230 nm
Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Formula Weight 18
Grade LC-MS
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 500 mL
Chemical Name or Material Water, CHROMASOLV™ LC-MS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.