missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™

Product Code. 16225301
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
4 x 4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™ LC-MS

  • Leading the industry for over 65 years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Synonym Aqua, Hydrogen Oxide, Hydrogen Hydroxide, Hydrate, Hydric Acid, Dihydrogen Monoxide
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade LC-MS
Packaging Glass Bottle
Quantity 2.5 L
Chemical Name or Material Water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'