missing translation for 'onlineSavingsMsg'
Learn More
Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™ Product Code.: 16419019

Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™

Product Code. 16419019
2.5 L
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
4 x 4 L
This item is not returnable. View return policy

Product Code. 16419019

Brand: Honeywell-Riedel-de Haen 392532.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™ LC-MS

  • Leading the industry for over 65 years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Synonym Aqua, Hydrogen Oxide, Hydrogen Hydroxide, Hydrate, Hydric Acid, Dihydrogen Monoxide
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade LC-MS
Packaging Glass Bottle
Quantity 2.5 L
Chemical Name or Material Water
Show More Show Less
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    26

    Hours

    20

    Minutes

    13

    Seconds

    36
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    57

    Hours

    20

    Minutes

    13

    Seconds

    36
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.