missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
59.06 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Specifications
pH | 6 to 8 (at 20°C) |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
UN Number | NONH for all modes of transport |
Beilstein | 2050024 |
Formula Weight | 18.02g/mol |
Grade | CHROMASOLV™ LC-MS Ultra |
Packaging | Glass Bottle |
Density | 1.000g/mL at 3.98°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'