missing translation for 'onlineSavingsMsg'
Learn More
Water, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals Product Code.: 10191231

Water, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Product Code. 10191231
1 L, Plastic Bottle
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10191231

Brand: Thermo Scientific Acros 327390010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, 100.0 deg.C, CHEBI:15377, 0.1ppm max., Undesignated, 1.0000g/mL, none detected, 16,383, 18.02, Molecular Biology, XLYOFNOQVPJJNP-UHFFFAOYSA-N, oxidane, 1ppb max., 15, 10237, MFCD00011332, 18.015, DNase, RNase, Protease free

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100.0°C
Linear Formula H2O
Fieser 16383
Merck Index 15, 10237
Formula Weight 18.02
Grade Molecular Biology
Packaging Plastic Bottle
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, DNase, RNase, Protease free
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465905 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number327390010,327390050
Lot NumberA0462102 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog Number327390010,327390050
Lot NumberA0462482 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number327390010,327390050
Lot NumberA0455073 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number327390010,327390050
Lot NumberA0440034 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number327390010,327390050
Product Identifier
  • Water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.