missing translation for 'onlineSavingsMsg'
Learn More
Water, Molecular Biology Grade, Fisher BioReagents Product Code.: 10505854
In Stock
to see stock.
Add to basket

Water, Molecular Biology Grade, Fisher BioReagents

Product Code. 10505854
1 L, Nalgene Poly Bottle
Click to view available options
Quantity:
100 mL
1 L
4 L
10 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
This item is not returnable. View return policy

Product Code. 10505854

Brand: Fisher BioReagents BP28191

In Stock
to see stock.
Edge
Add to basket
Add to basket
Request Bulk or Custom Formats
This item is not returnable. View return policy

For many fundamental molecular biology procedures

Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity <2μS/cm
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Grade DNase-, RNase- and Protease-Free
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.3μ filter
Treatment(s) Deionized
Density 1.000g/cm³
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, 0.03μm filtered to ensure high purity
Show More Show Less
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, Molecular Biology Grade, Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.