missing translation for 'onlineSavingsMsg'
Learn More

Water, Nuclease-free, Molecular Biology Grade, Ultrapure

Product Code. 15815408 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
1 L
5 L
Packaging:
Nalgene Bottle
Upright carton
Unit Size:
1 L
100mL
10mL
5 L
500mL
This item is not returnable. View return policy

Product Code. 15815408

Brand: Thermo Scientific J71786.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Bottle
Density 1
Quantity 100 mL
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.