missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Nuclease-free, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
1 L
5 L
Packaging:
Nalgene Bottle
Upright carton
Specifications
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
Formula Weight | 18.02 |
Grade | Molecular Biology |
Packaging | Nalgene Bottle |
Density | 1 |
Quantity | 1 L |
Specific Gravity | 1.000 |
Physical Form | Liquid |
Chemical Name or Material | Water |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
240339 | Certificate of Analysis | 11/06/2024 | J71786.X^,J71786.K2,J71786AE,J71786.K8,J71786.AP |
241531 | Certificate of Analysis | 11/06/2024 | J71786.X^,J71786.K2,J71786AE,J71786.K8,J71786.AP |
235131 | Certificate of Analysis | 15/02/2024 | J71786.X^,J71786.K2,J71786AE,J71786.K8,J71786.AP |
197595 | Certificate of Analysis | 15/11/2023 | J71786.X^,J71786.K2,J71786AE,J71786.K8,J71786.AP |
203204 | Certificate of Analysis | 13/11/2023 | J71786.X^,J71786.K2,J71786AE,J71786.K8,J71786.AP |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water, Nuclease-free, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction