missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Optima™ LC/MS Grade, Fisher Chemical™
28.17 GBP valid until 2024-07-31
Use promo code "21812" to get your promotional price.
Use promo code "21812" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Description
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Absorbance | ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm |
pH | 7 |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Merck Index | 15, 10237 |
Formula Weight | 18.02 |
Grade | LC-MS & UHPLC-UV |
Packaging | Glass Bottle |
Residue after Evaporation | ≤1ppm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'