Learn More
Water, pure, demineralized, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
pH | 6 to 8 |
Color | White |
Boiling Point | 100.0°C |
Conductivity | 5.0μS/cm max. |
Linear Formula | H2O |
Fieser | 16,383 |
Merck Index | 15, 10237 |
Formula Weight | 18.02 |
Grade | Pure |
Packaging | Plastic Drum |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467000 | Certificate of Analysis | 01/11/2024 | 276010200,276010050 |
A0457204 | Certificate of Analysis | 20/11/2023 | 276010200,276010050 |
A0457205 | Certificate of Analysis | 03/11/2023 | 276010200,276010050 |
A0452784 | Certificate of Analysis | 25/08/2023 | 276010200,276010050 |
A0452783 | Certificate of Analysis | 25/08/2023 | 276010200,276010050 |
Safety and Handling
- Water
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
0Minutes
35Seconds
36 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
23Minutes
24Seconds
23
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.