missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Water purification

Wall fitting E 313 for the manual production of demineralised water

Brand:  Miele Professional™ 3830330

Product Code. 10089260

  • £301.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pressurised to 10 bar
  • Complete with pressure hose 150 cm
Specifications

Specifications

Black
Wall fitting
Plastic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers