Learn More
Elgas™ Boost Pump, PURELAB flex 3 and 4 Water Purification System
Elgas™ Boost Pump, PURELAB flex 3 and 4 Water Purification System increases water pressure to facilitate flow and processing rate where necessary.
Brand: Elgas™ PUDPNY0150
This item is not returnable.
View return policy
Description
- Increases water pressure to boost processing and flow rate
- Suitable for PURELAB flex 3 and 4 water purification systems
Specifications
Increasing water pressure | |
PURELAB flex 3 and 4 water purification systems | |
3 kg |