Learn More
Elgas™ Regulator, PURELAB flex 3 and 4 Water Purification System
Elgas™ Regulator, PURELAB flex 3 and 4 Water Purification System can be used to moderate feedwater pressure where it exceeds specified limits.
Brand:  Elgas™ VAPSAC0003
Product Code. 13165980
  • £425.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moderates water pressure to appropriate system capacity
Specifications
Specifications
Moderating water pressure
PURELAB flex 3 and 4 water purification systems