Learn More
Elgas™ PURELAB flex 4 Water Purification System
Elgas™ PURELAB flex 4 Water Purification System is an independent system with manual filling capacity that can operate independently from a tap.
Brand:  Elgas™ PRIPLB0163
Product Code. 13145970
  • £10635.00 / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ultrapure Type I water
  • Can be connected to a pre-treated water supply and is particularly suited for small volumes of water where TOC levels are critical and must remain stable.
  • Space saving design.
    • The compact unit can be placed on the bench or can be wall mounted and has an integrated 7 liter reservoir, ensuring that water is always available.
  • Portable system.
    • Access on the top of the system gives the option to fill the reservoir with pre-purified water. This means that it can be moved at any time as the lab environment evolves.
  • Real-time TOC Monitoring.
    • Provides complete confidence in organic purity and clear display at all times.
  • Flexible dispenser.
    • The intuitive dispenser offers a clear display of the water purity for absolute confidence as you dispense.
  • Easy to maintain.
    • The front doors ensure easy access to the consumables, making them quick and easy to replace.
  • Data capture.
    • Data capture via USB for system performance validation and software updates.
Specifications
Specifications
Daily volume: <100 L
900 to 1020 mm
23 kg
Up to 2.0 L/min.
470 x 236 x 900-1020 mm
470 mm
236 mm