Learn More
Elgas™ Reverse Osmosis Module Purelab flex 3 Water Purification System
Elgas™ Reverse Osmosis Module for PURELAB flex 3 Water Purification System is a semipermeable membrane to remove ions, molecules, and larger particles in the PURELAB flex 3, a small but powerful tap-to-ultrapure water production system.
Brand:  Elgas™ MBROMD0031
Product Code. 13185980
  • £378.00 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Semipermeable membrane to remove ions, molecules, and larger particles from drinking water
  • Suitable for PURELAB flex 3 water purification system
Specifications
Specifications
10 L/hr.
Water purification
PURELAB flex 3 system