missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water Purification Systems Storage Reservoirs

Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.

Brand:  Thermo Scientific™ 50155494

 View more versions of this product

Product Code. 11367081

  • £1585.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.

Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 L
Storage Reservoir, with level display and recirculation pump
Pacific TII
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers