missing translation for 'onlineSavingsMsg'
Learn More

Water, for qPCR or RT-qPCR, Fisher BioReagents

Product Code. 15812544
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
10 L
Packaging:
Poly Bottle
Poly CUBE
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity water rigorously tested for degrading enzymes and genomic DNA that interefere with amplification and can lead to false-positive signals in PCR, qPCR and RT-qPCR applications.

  • Minimize the risk of false positive signals
  • Tested for DNase, RNase, protease, nickase, and E. coli and human genomic DNA using sensitive assays
  • Evaluated for metal impurities at low ppb concentrations to ensure optimal enzymatic activity, especially for polymerase during PCR
  • Sub-micron filtered to remove particulates
  • Suitable for use in qPCR and RT-qPCR for genomics work, microarrays, and other sensitive life science applications
  • Also useful for the manufacturing of PCR-related kits and mastermixes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 5.4 to 7.0
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity ≤2μS/cm
DNase Not detected
Endotoxin <0.1EU/mL
Formula Weight 18.02
Grade qPCR
Packaging Poly Bottle
Quantity 1 L
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'