Learn More
Water R Solution, Fisher Chemical™ Product Code.: 12940564
Estimated Shipment: 10-03-2025
to see stock.
Add to basket
Water R Solution, Fisher Chemical™
Product Code. 12940564
5000 mL
Click to view available options
Quantity:
5000 mL
Product Code. 12940564
Brand: Fisher Chemical RP 226 5000
Estimated Shipment: 10-03-2025
to see stock.
Add to basket
Add to basket
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 5000 mL
Physical Form Liquid
Product Content Correction
Product Title
Water R Solution, Fisher Chemical™ >