missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™
TraceSELECT™, for trace analysis
£20.99 - £329.88
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627550
|
Honeywell-Riedel-de Haen
95305-250ML |
250 mL | Poly Bottle |
£20.99
250mL |
In Stock
Log in to see stock. |
||||
15647550
|
Honeywell-Riedel-de Haen
95305-500ML |
500 mL | Poly Bottle |
£23.57
500mL |
In Stock
Log in to see stock. |
||||
15607550
|
Honeywell-Riedel-de Haen
95305-1L |
1 L | Glass Bottle |
£49.89
1 L |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
15617550
|
Honeywell-Riedel-de Haen
95305-2.5L |
2.5 L | Poly Bottle |
£92.22
2.50 L |
In Stock
Log in to see stock. |
||||
15697540
|
Honeywell-Riedel-de Haen
95305-10L |
10 L | Cubitainer |
£120.62
10 L |
In Stock
Log in to see stock. |
||||
15637550
|
Honeywell-Riedel-de Haen
95305-4X2.5L |
4 x 2.5 L | Poly Bottle |
£329.88
Pack of 4 |
Estimated Shipment: 11-11-2024
Log in to see stock. |
||||
Description
High purity solvents for speciation analysis by LC-ICP-MS
Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance
The blank values for metal traces in these solvents are in the ppb range or lower.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
6 to 8 (at 20°C) | |
Colorless | |
100°C | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
Odorless | |
Liquid |
7732-18-5 | |
0°C | |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
TraceSELECT™ | |
1g/mL at 20°C | |
Water |
Safety and Handling
missing translation for 'rtecsNumber' : 231-791-2