missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Water, UHPLC-MS, Thermo Scientific™

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.

Brand:  Thermo Scientific W8-1

72.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15339865

  • £123.00 / 1 L
In Stock
to see stock.
Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.01 AU at 230nm, <-0.01 AU at 210nm, <-0.01 AU at 220nm, <-0.01 AU at 240nm, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, 2ppb max., 100 deg.C, 5ppb max., CHEBI:15377, Colorless, 1.000g/cm3, UHPLC-MS

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
7
Colorless
100°C
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Clear Borosilicate Glass Bottle
17.5mmHg at 20°C
1 L
Water
7732-18-5
0°C
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
UHPLC-MS
≤1ppm
1.000 g/cm3
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number245790 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberW8-1
Lot Number245935 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberW8-1
Lot Number241962 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberW8-1
Lot Number200034 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberW8-1
Lot Number200849 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberW8-1
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Water, UHPLC-MS, Thermo Scientific™ > Clear Borosilicate Glass Bottle; 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only