missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
LDPE Bottle
Unit Size:
1 L
2 L
500mL
Description
- Evaluated for 65 metal impurities in ppt range
- Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
- Packaged in precleaned LDPE bottles to maintain product integrity
Specifications
Specifications
| pH | 7 |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Formula Weight | 18.02 |
| Grade | Ultra Trace Elemental Analysis |
| Packaging | LDPE Bottle |
| Vapor Pressure | 17.5mmHg at 20°C |
| Density | 1.000g/cmÂł |
| Certificate of Analysis | Certificate of Lot Analysis included with each bottle. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction
