Learn More
Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals
£35.58 - £221.52
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348217
|
Thermo Scientific Alfa Aesar
022934.K2 |
1 L |
£35.58
1 L |
In Stock
Log in to see stock. |
|||||
11338217
|
Thermo Scientific Alfa Aesar
022934.K7 |
4 L |
£65.53
4 L |
In Stock
Log in to see stock. |
|||||
11358217
|
Thermo Scientific Alfa Aesar
022934.M6 |
4 x 4 L | N/A | ||||||
Description
Water ultrapure is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is also used in conductometric titration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
100°C | |
MFCD00011332 | |
Miscible with methanol,acetone,ethanol. | |
O | |
18.015 | |
CHEBI:15377 | |
HPLC | |
1.333 | |
Water |
0°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
1.000 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 231-791-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.