missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with0.1% Formic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
CAS | 7732-18-5, 64-18-6 |
Quantity | 2 L |
Packaging | Glass Bottle |
Molecular Formula | H2O |
MDL Number | MFCD00011332 |
UN Number | NONH for all modes of transport |
Formula Weight | 18.02g/mol |
Grade | LC-MS Ultra CHROMASOLV™ |
Chemical Name or Material | Water With 0.1% Formic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water with0.1% Formic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™ >
Spot an opportunity for improvement?Share a Content Correction