Learn More
Thermo Scientific™ WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™
Use in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult™ format.
Brand: Thermo Scientific™ R4757090
Code : 65
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Description
Thermo Scientific™ WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™ is used in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
- Deliver <50 CFUs in a single inoculum to mimic water samples
- One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
Specifications
13047TM*, 13047™* | |
Quanti-Cult | |
Gram-negative Rods | |
10 Test/Kit |
Enterobacter cloacae subsp. cloacae ATCC™ 13047™ | |
Quality Control Organism | |
Quantitative QC | |
10 Tests/Kit |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.