missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WaterBugs™ Klebsiella pneumoniae Strain C6

Product Code. 12996047
Click to view available options
Quantity:
10 Tests/Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12996047

Brand: Thermo Scientific™ R4753935

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.

Thermo Scientific™ WaterBugs Klebsiella pneumoniae Strain C6 is used for water/wastewater quality control testing; microorganisms are supplied in a Quanti-Cult format.

  • Deliver <50 CFUs in a single inoculum to mimic water samples
  • One test/reconstituted vial

Ordering Alerts:

Supplied with 10 preserved organism vials; 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

Description Klebsiella pneumoniae Strain C6
Format Quanti-Cult
Product Type Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Quantity 10 Tests/Kit

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.