Learn More
Thermo Scientific™ WaterBugs™ Pseudomonas aeruginosa ATCC™ 27853™
Use in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult™ format.
Brand:  Thermo Scientific™ R4757060
Product Code. 12916057
  • £596.00 / Each
Explore more special offers
Description
Description
Thermo Scientific™ WaterBugs™ Pseudomonas aeruginosa ATCC™ 27853™ is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • Deliver <50 CFUs in a single inoculum to mimic water samples
  • One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
27853TM*, 27853™*
Quanti-Cult
Gram-negative Rods
10 Test/Set
Boston 41501
Pseudomonas aeruginosa ATCC™ 27853™
Quality Control Organism
Quantitative QC
10 Test/Set
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.