Learn More
Emma Safety Footwear Waterspot Spray
Waterspot spray.
Brand: Emma Safety Footwear MM00005600
Additional Details : Weight : 0.50000kg Transport : UN number : 1950 Chem class : 2.1
Description
Waterspot spraySpecifications
400mL | |
Waterspot spray |
Safety and Handling
- Schuhspray (VE=400 ml)
Signal Word
- Danger
Hazard Category
- Flammable and non-flammable aerosols category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H222-Extremely flammable aerosol.
- H229-Pressurised container: May burst if heated.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P211-Do not spray on an open flame or other ignition source.
- P251-Do not pierce or burn, even after use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P391-Collect spillage.
- P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isobutane, pentane,propane