Learn More
Watson-Marlow Marprene Double Segment Manifold Tubing
Brand: Watson-Marlow 979.0025.000
Additional Details : Weight : 0.06000kg
Description
- Made from autoclavable marprene
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
Manifold Tubing | |
Marprene | |
0.25 mm |
0.25 mm | |
400DM2 and 400DM3 pumpheads |