Learn More
Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Brand:  Watson-Marlow 979.0114.000
 View more versions of this product
Product Code. 17847953
  • £289.00 / Pack of 6
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Double Segment Tubing
1.14 mm
0.03 in.
323S/MC peristaltic pump
0.12 to 8.1 (four roller pump), 0.09 to 6.6 (eight roller pump)mL/min.
1/25 in.
Marprene
0.8 mm
6 Pk.
Red/Red