missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ WAVE Maxi with Caster and Additional Casting Unit

Novel vertical screw-clamp technology reduces the number of screws required for set up, dramatically reducing assembly time. A built-in inner buffer chamber within the PAGE insert allows set up to be completed without inclusion of a top tank or upper buffer chamber.

Brand:  Fisherbrand™ FVS20WAVESYS-CU

Product Code. 12633546

  • £1105.00 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed to perform a variety of separations, including first and second-dimension SDS-PAGE, native, preparative, gradient and high-resolution nucleic acid electrophoresis, plus capillary tube gel IEF and electroblotting
  • Innovative, vertical screw-clamp technology within the PAGE insert
  • Only four screws are now required to secure as many 20 x 20 cm gels
  • Much faster set up speed compared to competitor products
  • Vertical screw-clamp configuration distributes pressure evenly along the height of the gel, rather than in the center to eliminate plate bowing and gel compression
  • Maintains a leak-proof seal during casting
  • Ergonomic, wave-like design of the PAGE insert aids with handling and set up
Specifications

Specifications

4 Gels
Casting Base, Additional Casting Unit
5300 mL
30 x 18 x 27 cm
16 x 17.5 cm
FVS20WAVESYSCU
1200 mL
2.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers