missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Wax Dispensers

Get on-demand delivery of molten wax with Epredia™ Wax Dispensers.

Brand:  Epredia™ A84600010

 View more versions of this product

Product Code. 15513646

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The inner tank is manufactured from aluminum for good thermal conductivity and improved temperature control. It has a filter screen fitted to prevent coarse particles from blocking the delivery tap. The tank is also fully insulated to prevent heat loss from the tank and to ensure that the outer surfaces of the tank are safe to touch.


Features
  • Space saving form factors
  • 7.5L or 15L capacity (6kg or 12kg of pelletized wax)
  • Non-drip, three-position lever tap–push dispense, lock open for rapid delivery, or lock closed for safety
  • High tap position (153mm from the bench) accommodates large containers for transferring molten wax
  • Integral tap filter screen (0.5mm mesh) prevents coarse particles from blocking the delivery tap
  • Anti-microbial coating (Ag+) inhibits growth of bacteria
Heating System
  • Ultra-fast heating system minimizes the wait for molten wax
  • Digital temperature control–high-capacity model features individual controls for each of the dual reservoirs
  • Aluminum inner tank provides good thermal conductivity and improved temp control
  • Insulated paraffin tank prevents heat loss from the tank and ensures that outer surfaces of the tank are safe to touch
  • Dedicated tap-heating system prevents blockages and ensures even flow of wax
Specifications

Specifications

15 L
230 V
High Capacity Wax Dispenser
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.