missing translation for 'onlineSavingsMsg'
Learn More

WAY 161503 hydrochloride, Tocris Bioscience™

Product Code. 30001453
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

Potent and selective 5-HT2C receptor agonist (Ki = 4 nM; EC50 = 12 nM). Antidepressant following systemic administration in vivo.

Chemical Identifiers

CAS 276695-22-8
Molecular Formula C11H12Cl3N3O
Molecular Weight (g/mol) 308.587
InChI Key YPNWSZJDAKOUAW-UHFFFAOYSA-N
PubChem CID 21976692
IUPAC Name 8,9-dichloro-1,2,3,4,4a,6-hexahydropyrazino[1,2-a]quinoxalin-5-one;hydrochloride
SMILES C1CN2C(CN1)C(=O)NC3=CC(=C(C=C32)Cl)Cl.Cl

Specifications

Quantity 10 mg
Synonym way 161503 hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h-pyrazino 1,2-a quinoxalin-5 6h-one hydrochloride, 8,9-dichloro-1h,2h,3h,4h,4ah,6h-pyrazino 1,2-a quinoxalin-5-one hydrochloride, way161503 hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h,6h-pyrazino 1,2-a quinoxalin-5-one hydrochloride, way 161503 hydrochloride hplc, 1h-pyrazino 1,2-a quinoxalin-5 6h-one, 8,9-dichloro-2,3,4,4a-tetrahydro, 8,9-dichloro-1h,2h,3h,4h,4ah,6h-piperazino 1,2-a quinoxalin-5-one hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h-pyrazino 1,2-a quinoxalin-5 6h-one-hydrogen chloride 1/1
Formula Weight 308.59
Percent Purity >99%
Chemical Name or Material WAY 161503 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'