missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAY 170523, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
Description
Potent and selective inhibitor of MMP-13 (IC50 values are 17, 945, > 1000 and > 10000 nM for MMP-13, MMP-9, TACE and MMP-1 respectively).
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 613.68 |
Percent Purity | >99% |
Chemical Name or Material | WAY 170523 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'