missing translation for 'onlineSavingsMsg'
Learn More

WAY 170523, Tocris Bioscience™

Product Code. 18065192
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective inhibitor of MMP-13

Potent and selective inhibitor of MMP-13 (IC50 values are 17, 945, > 1000 and > 10000 nM for MMP-13, MMP-9, TACE and MMP-1 respectively).

Chemical Identifiers

CAS 307002-73-9
Molecular Formula C33H31N3O7S
Molecular Weight (g/mol) 613.685
InChI Key FARMEEAGJWMFSZ-UHFFFAOYSA-N
Synonym n-2-4-benzyl 2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-carboxamide, d0ll8i, n-2-4-2-hydroxyamino carbonyl-4,6-dimethylphenyl phenylmethyl amino sulfonyl phenoxy ethyl-2-benzofurancarboxamide, way hplc, benzofuran-2-carboxylic acid 2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethyl-phenyl-sulfamoyl-phenoxy-ethyl-amide, n-2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-car
PubChem CID 9830392
IUPAC Name N-[2-[4-[benzyl-[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl]phenoxy]ethyl]-1-benzofuran-2-carboxamide
SMILES CC1=CC(=C(C(=C1)C(=O)NO)N(CC2=CC=CC=C2)S(=O)(=O)C3=CC=C(C=C3)OCCNC(=O)C4=CC5=CC=CC=C5O4)C

Specifications

Quantity 10 mg
Formula Weight 613.68
Percent Purity >99%
Chemical Name or Material WAY 170523
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'