missing translation for 'onlineSavingsMsg'
Learn More
WEB 2086, Tocris Bioscience™ Product Code.: 18782784

WEB 2086, Tocris Bioscience™

Product Code. 18782784
10 mg
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy

Product Code. 18782784

Brand: Tocris Bioscience 2339/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PAF receptor antagonist

Potent, selective platelet-activating factor (PAF) receptor antagonist (Ki = 16.3 nM). Displays anti-inflammatory, antiangiogenic and anticancer activity. Inhibits growth and proliferation of MCF-7 breast cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105219-56-5
Molecular Formula C22H22ClN5O2S
Molecular Weight (g/mol) 455.961
InChI Key JGPJQFOROWSRRS-UHFFFAOYSA-N
Synonym apafant, apafanto, apafantum, triazolodiazepine, apafant usan:inn, apafantum inn-latin, apafanto inn-spanish, web 2086bs, unii-j613ni05sv, 3h apafant
PubChem CID 65889
SMILES CC1=NN=C2N1C3=C(C=C(S3)CCC(=O)N4CCOCC4)C(=NC2)C5=CC=CC=C5Cl

Specifications

Quantity 10 mg
Formula Weight 455.96
Percent Purity >99%
Chemical Name or Material WEB 2086
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
WEB 2086, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.