missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Individual Closures

Save time and minimize the risk of vial contamination prior to use with 96-Well Plate Kits packaged with vials sealed with individual closures.

Brand:  Thermo Scientific™ 60180-K204

 View more versions of this product

Product Code. 15142799

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Save time and minimize the risk of vial contamination prior to use with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Individual Closures. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates and glass vials with pre-attached caps and septa.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Webseal
Glass Inserted Plate Kit with Individual Closures
96
1000 μL
Vial: 60180-V111; Cap: 60180-C101
Round
40 mm
96 Micro-Tube-Rack-System with PE Plug Septa
1 Pk.
Clear
Glass, Polyethylene
800 μL
Round
1
1/Pk.
7.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers