missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WebSeal™ Sealing Tapes

Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.

Brand:  Thermo Scientific™ 60180-M142

328.74 GBP valid until 2025-12-31
Use promo code "24292" to get your promotional price.


 View more versions of this product

Product Code. 15122719

  • £377.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.

Easy, Consistent Sealing

  • Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Tapes seal onto the plate with a convenient, handheld applicator (available separately)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polyethylene, Silicone
Non-sterile Sealing Tape
Pressure Sensitive
All Microplates
Sealing Tape, -40°C - 90°C
100
Webseal
-40°C to +90°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ WebSeal™ Sealing Tapes > PE/Silicone

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.