missing translation for 'onlineSavingsMsg'
Learn More

Salter Balance 1,000g spring flat case x 10g

Brand:  Salter SAFN00500010000

Balance 1,000g spring flat case x 10g

Code : 03

Additional Details : Weight : 0.09024kg

Product Code. 12767046

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers