missing translation for 'onlineSavingsMsg'
Learn More

Clark Scientific™ Weighing Boat

Brand:  Clark Scientific™ ADS30W

Additional Details : Weight : 0.41400kg

 View more versions of this product

Product Code. 10369680

  • £27.05 / Pack of 500
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

30 mL
Diamond
55 x 85 mm
85 mm
White
Weighing Boat
55 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers