missing translation for 'onlineSavingsMsg'
Learn More

Clark Scientific™ Weighing Boat

Brand:  Clark Scientific™ DS5W

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10450961

  • £29.85 / Pack of 1000
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

5 mL
Rhombus
31 x 54 mm
54 mm
White
Weighing Boat
31 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers