missing translation for 'onlineSavingsMsg'
Learn More

Clark Scientific™ Weighing Boat

Brand:  Clark Scientific™ ASQ250W

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10617491

  • £122.81 / Pack of 1000
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

250 mL
Square
134 x 134 mm
134 mm
White
Weighing Boat
134 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers