missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Weighing Bottles with NS Stopper

Product Code. 10028830
Click to view available options
Capacity (Metric):
5 mL
70 mL
80 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • With interchangeable NS stopper
  • With inscription label, with fused-in size (mm) and capacity (ml)
  • Available in tall or low form
  • Stopper: DURAN™, bottle made of DURAN™ tubing

Specifications

Capacity (Metric) 70 mL
Material Glass
Height (Metric) 80 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'