missing translation for 'onlineSavingsMsg'
Learn More

Block Technology Weighing Isolator and Containment Isolator

Isolator suitable for work with API/HAPI and for protection of personnel. This isolator is designed for work with hazardous materials and is used for personnel protection. The isolator can be used for weighing, sampling and handling material, and as an isolator in a pharmacy.

Brand:  Block Technology IZVW


Additional Details : Weight : 690.00000kg

Product Code. 17351689

  • POA / Each
Estimated Shipment: 04-12-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Dual-chamber weighing isolator
  • Airlock with one glove allowing for loading of material and instruments
  • Working chamber with three gloves intended for work with hazardous materials
  • Possible to ensure inert atmosphere inside chambers (N2, Ar)
  • Leak tightness class 3 according to ISO 10648-2
  • 'C' Class cleanliness according to EU GMP standard
  • 'Audit Trail' compliant
  • Electronic control system for automatic adjustment of basic operating modes managed by Siemens PLC
  • Color touchscreen controls
  • Jacket material: stainless steel AISI 304
  • Isolator working chamber material – AISI 316L with a thickness of 3.00 mm
  • Polished surface finish, Ra < 0.6 μm
  • Inlet filtration HEPA H14 - 'C' Class cleanliness
  • Safe filtration of exhaust air with double HEPA H14 filters
  • Easy-to-clean inner and outer surfaces
  • Sliding trays between chambers for easy material transfer
  • Oval holes with gloves for improved material handling
  • Turbulent flow
  • Negative pressure mode

Turbulent Flow - Commonly used for 'C' and 'D' classes of cleanliness where the direction of airflow is not clearly defined. The air flows in an irregular direction from the air supply inlet to the air outlet through cylindrical HEPA filters H14 at a filtering efficiency of 99.995%. The rate of air exchanges of clean air into the chamber usually ranges from 5 to 40 exchanges per hour.

The HVAC system allows for the formation of negative pressure in the interior of the isolator. This function is mostly used in case of a failure – compromised leak-proofness - and helps to protect personnel. Another level of protection for critical points in the process is the creation of a pressure cascade so that the location of dangerous substance handling contains the highest level of negative pressure. This minimizes the risk of contaminating other chambers or the surroundings. In order to prevent damage to the equipment caused by insufficient pressure a pneumatic valve is used. This protective function prevents the pressure from dropping further, thus protecting the technology from damage.

Standards

  • Cable glands - Cable glands are mounted in the isolator wall.
  • Sockets for additional equipment power supply - Sockets are placed on the isolator rear wall. Sockets are operable from the control display. Sockets available in all country variants.
  • Wire trays & accessories - The rear panel can be fitted with wire trays and hangers made of AISI 316L stainless steel.
  • Built-in LED lights - Built-in LED lights ensure illumination levels of at least of 500 lux.

Safety Features

  • Pressure fuse - The pressure fuse serves as a safety element indicating increased pressure in the chamber. In the event that the assigned pressure in the chamber is exceeded a pneumatic flap valve is opened and the pressure value is decreased without deterioration of internal atmosphere.
  • Safety glass - We use tempered safety glass for the front windows and ports of our equipment. This glass has improved mechanical properties and is durable. When damaged it forms small chunks which reduce the risk of injury from cuts.
  • Safe replacement of sleeves - Our glove / sleeve flanges have two grooves. The first for sealing the glove and the second for an o-ring. This design allows a standardised procedure for the safe replacement of gloves.
  • Independent UPS power supply - An independent UPS (Uninterruptible Power Supply) can be installed inside the isolator. In the event of an interruption in the supply of electricity, short-term operation is maintained through a power supply in the isolator to allow the process and the environment to be secured.
  • Safe replacement of the main filter
Specifications

Specifications

Weighing/Containment Isolator
1000 mm
1900 mm
Stainless Steel (304), Stainless Steel (316L)
2550 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers