missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Weighing Pan
Weighing Pan accessory for Cubis™ II
£240.00
Specifications
For Use With (Equipment) | Cubis™ II High-Capacity Micro Balance |
---|---|
Type | Weighing Pan |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17997320
|
Sartorius
YWP09-3 |
50 mm |
£240.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17938240
|
Sartorius
YWP10-3 |
90 mm |
£240.00
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Cubis™ II Accessory
Specifications
Cubis™ II High-Capacity Micro Balance | |
Weighing Pan |